Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHMP7 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318469AF350
Description
CHMP7 Polyclonal antibody specifically detects CHMP7 in Human samples. It is validated for Western BlotSpecifications
CHMP7 | |
Polyclonal | |
Western Blot | |
charged multivesicular body protein 7, CHMP family, member 7, Chromatin-modifying protein 7, MGC29816 | |
Recognizes region between residues Between Between 400 and 450.. | |
0.1 mL | |
Autophagy, Cell Biology, Signal Transduction | |
91782 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction