Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHMP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116579 Supplier Thermo Scientific Supplier No. PA5116579
Add to Cart
Add to Cart

CHMP7 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CHMP7
Gene Accession No. Q8R1T1, Q8WUX9
Gene Alias 4930596K11Rik; 6330407G04Rik; AI450338; AW550775; Charged multivesicular body protein 7; CHMP family, member 7; Chmp7; Chromatin-modifying protein 7; MGC29816; RGD1308779
Gene Symbols CHMP7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 230-453 of human CHMP7 (NP_6894851).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105513, 364419, 91782
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.