Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHMP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109533
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109533 Supplier Invitrogen™ Supplier No. PA5109533
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQKEKRVTVL EQNGEKIVKF ARGPRAKVSP VNDVDVGVYQ LMQSEQLLSR KVESLSQEAE RCKEEARRAC RAGKKQLALR SLKAKQRTEK RIEALHAKLD TVQGILDRIY ASQTDQMVFN AYQAGVGALK LSMKDVTVEK AESLVDQIQE LCDTQDEVSQ TLAGGVTNGL DFDSEELEKE LDILLQDTTK EPLDLPDNPR NRHFTNSVPN PRISDAELEA ELEKLSLSEG GLVPSSKSPK RQLEPTLKPL.

CHMP7 gene ontology annotations related to this gene include vacuolar transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CHMP7
Gene Accession No. Q8R1T1
Gene Alias 4930596K11Rik; 6330407G04Rik; AI450338; AW550775; Charged multivesicular body protein 7; CHMP family, member 7; Chmp7; Chromatin-modifying protein 7; MGC29816; RGD1308779
Gene Symbols CHMP7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 204-453 of human CHMP7 (NP_689485.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105513
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.