Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHN1 Monoclonal Antibody (OTI3C6)
GREENER_CHOICE

Catalog No. PIMA526053
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526053 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526053 Supplier Invitrogen™ Supplier No. MA526053
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CHN1 Monoclonal Antibody for Western Blot, IHC (P)

GTPase-activating protein for p21-rac and a phorbol ester receptor. Involved in the assembly of neuronal locomotor circuits as a direct effector of EPHA4 in axon guidance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Chn1
Gene Accession No. P15882, P30337, Q91V57
Gene Alias 0610007I19Rik; 0710001E19Rik; 1700112L09Rik; 2900046J01Rik; a2-chimaerin; A-chimaerin; AI413815; alpha1 chimaerin; alpha2 chimaerin; alpha3-chimerin; alpha-chimerin; ARHGAP2; chimaerin; chimaerin 1; chimerin (chimaerin) 1; chimerin 1; CHN; CHN1; DURS2; NC; N-chimaerin; N-chimerin; rho GTPase-activating protein 2; RHOGAP2
Gene Symbols Chn1
Host Species Mouse
Immunogen Full length human recombinant protein of CHN1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108699, 1123, 478805, 84030
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.