Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121116
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121116 Supplier Invitrogen™ Supplier No. PA5121116
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: BT-474, SH-SY5Y, Jurkat, HeLa, MCF7, Mouse testis, Mouse brain, Mouse spinal cord, Rat brain. Immunogen sequence: MALTLFDTDE YRPPVWKSYL YQLQQEAPHP RRITCTCEVE NRPKYYGREF HGMISREAAD QLLIVAEGSY LIRESQRQPG TYTLALRFGS QTRNFRLYYD GKHFVGEKRF ESIHDLVTDG LITLYIETKA AEYIAKMTIN PIYEHVGYTT LNREPAYKKH MPVLKETHDE RDSTGQDGVS EKRLTSLVRR ATLKENEQIP.

GTPase-activating protein for p21-rac and a phorbol ester receptor. Involved in the assembly of neuronal locomotor circuits as a direct effector of EPHA4 in axon guidance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Chn1
Gene Accession No. P15882, P30337, Q91V57
Gene Alias 0610007I19Rik; 0710001E19Rik; 1700112L09Rik; 2900046J01Rik; a2-chimaerin; A-chimaerin; AI413815; alpha1 chimaerin; alpha2 chimaerin; alpha3-chimerin; alpha-chimerin; ARHGAP2; chimaerin; chimaerin 1; chimerin (chimaerin) 1; chimerin 1; CHN; CHN1; DURS2; NC; N-chimaerin; N-chimerin; rho GTPase-activating protein 2; RHOGAP2
Gene Symbols Chn1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human CHN1 (NP_001813.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108699, 1123, 84030
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.