Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHN48 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598366
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598366 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598366 Supplier Invitrogen™ Supplier No. PA598366
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHN48 Polyclonal Antibody for Western Blot, ELISA

CHN48 is involved in defense against chitin-containing fungal pathogens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHN48
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Host Species Rabbit
Immunogen Recombinant Nicotiana tabacum Endochitinase A protein (24-322AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.