Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ CHO CD EfficientFeed™ A AGT™ Nutrient Supplement
Click to view available options
Reconstituted Volume:
10 L
100 L
Description
Gibco™ CHO CD EfficientFeed™ A nutrient supplement is an animal origin-free, chemically defined complete feed formulation that provides up to 2-5 fold improvement in titers used in fed batch versus batch culture. It contains no proteins, hydrolysates, or components of incompletely defined composition. CHO CD EfficientFeed™ A AGT™ (Advanced Granulation Technology™) Kit Nutrient Supplement provides all the benefits of CHO CD EfficientFeed™ A liquid supplement in a simpler to scale up and reconstitute 2-part kit that is primarily AGT™ dry format material.
The AGT™ format is designed for large scale manufacturing and ease of use. This product is also available in liquid form for small scale use and experimentation.
Expect up to 2- to 5-fold increases in productivity
Avoid issues associated with the use of hydrolysates and other alternatives while gaining superior performance
Simplify scale-up by leveraging the value of AGT™ simple reconstitution
Achieve performance comparable to original liquid supplement
Ready for larger scale use with convenient packaging options (10 L and 100 L)
Which EfficientFeed™ Nutrient Supplement to Choose?
Consider the CHO CD Efficient Feed™ Kit when:
You are using a base medium with less than 6 g/L glucose and no hydrolysate at day 0
Consider CHO CD EfficientFeed™ A supplement when:
You are currently using CD OptiCHO™ as your base cell growth medium
Consider CHO CD EfficientFeed™ B supplement when:
You are currently using CD CHO as your base cell growth medium
Consider CD EfficientFeed™ C AGT™ supplement when:
You want higher productivity in GS CHO and other selected cell lines
You are currently using CD FortiCHO™ as your base cell growth medium
You are using a base medium with greater than 6 g/L glucose or a hydrolysate at day 0
Increased Titers
CHO CD EfficientFeed™ supplements have been shown to increase titers 2-5 fold versus batch culture, and are an excellent starting point for process development (Figures 1a & 1b). Furthermore, they outperform competitive alternatives, including less consistent hydrolysates (Figures 2, 3a, & 3b).
Simple Scale-Up and Cost Savings
The ability to scale up in a dry format provides ease of handling and storage in manufacturing and other bioprocess settings. With AGT™ (Advanced Granulation Technology™) dry format, it is possible to store and use the product even more easily and safely than traditional dry format products. CHO CD EfficientFeed™ A AGT™ supplement is offered in 10 L and 100 L packaging that allows scale-up from the 1 L liquid supplement as a seamless process, as well as allow for cost savings when purchasing production level volumes.
Comparable Performance to Liquid Supplement
Equally important as the convenience and simplicity of AGT™ (Advanced Granulation Technology™) dry format is the comparable performance to the liquid CHO CD EfficientFeed™ A version (Figure 4). Both forms have performed similarly in our testing, and the AGT™ version has been used successfully as a prototype in many processes by multiple companies.
AGT™ Format Ease of Use
CHO CD EfficientFeed™ A AGT™ Kit Nutrient Supplement delivers dry format scale-up convenience as well as simple reconstitution with approximately 1% concentrated solution add back. For more details, consult the supporting documents provided below.
For research or further manufacturing use. Not for use in diagnostic procedures.
The AGT™ format is designed for large scale manufacturing and ease of use. This product is also available in liquid form for small scale use and experimentation.
Which EfficientFeed™ Nutrient Supplement to Choose?
Consider the CHO CD Efficient Feed™ Kit when:
Consider CHO CD EfficientFeed™ A supplement when:
Consider CHO CD EfficientFeed™ B supplement when:
Consider CD EfficientFeed™ C AGT™ supplement when:
Increased Titers
CHO CD EfficientFeed™ supplements have been shown to increase titers 2-5 fold versus batch culture, and are an excellent starting point for process development (Figures 1a & 1b). Furthermore, they outperform competitive alternatives, including less consistent hydrolysates (Figures 2, 3a, & 3b).
Simple Scale-Up and Cost Savings
The ability to scale up in a dry format provides ease of handling and storage in manufacturing and other bioprocess settings. With AGT™ (Advanced Granulation Technology™) dry format, it is possible to store and use the product even more easily and safely than traditional dry format products. CHO CD EfficientFeed™ A AGT™ supplement is offered in 10 L and 100 L packaging that allows scale-up from the 1 L liquid supplement as a seamless process, as well as allow for cost savings when purchasing production level volumes.
Comparable Performance to Liquid Supplement
Equally important as the convenience and simplicity of AGT™ (Advanced Granulation Technology™) dry format is the comparable performance to the liquid CHO CD EfficientFeed™ A version (Figure 4). Both forms have performed similarly in our testing, and the AGT™ version has been used successfully as a prototype in many processes by multiple companies.
AGT™ Format Ease of Use
CHO CD EfficientFeed™ A AGT™ Kit Nutrient Supplement delivers dry format scale-up convenience as well as simple reconstitution with approximately 1% concentrated solution add back. For more details, consult the supporting documents provided below.
For research or further manufacturing use. Not for use in diagnostic procedures.
Specifications
Specifications
Content And Storage | Store at 2 to 8°C |
Cell Line | CHO |
Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
Culture Type | Suspension Cell Culture |
Form | AGT |
Product Type | Nutrient Supplement |
Serum Level | Serum-Free |
Sterility | Non-sterile |
With Additives | Sodium Pyruvate, Sodium Bicarbonate |
Without Additives | No Glutamine, No Phenol Red, No Insulin |
Show More |
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction