Learn More
CHO cells Rabbit anti-Hamster, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 278031AP150UL

Description
This antibody is raised by the total CHO cell lysate. It detects all the CHO proteins.
CHO cells (Chinese Hamster Ovary cells) are a laboratory-cultured cell line derived from cells of the ovaries of Chinese hamsters.Specifications
CHO cells | |
Polyclonal | |
Unconjugated | |
Chinese Hamster Ovary cells, CHO, CHO cells | |
Antigen Affinity Chromatography | |
RUO | |
Hamster | |
Antibody | |
IgG |
Western Blot | |
1.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Rabbit | |
150 μL | |
Primary | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.