Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cholecystokinin-B R/CCKBR Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB1002803
Description
Cholecystokinin-B R/CCKBR Polyclonal specifically detects Cholecystokinin-B R/CCKBR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
Cholecystokinin-B R/CCKBR | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
CCK2 receptor, CCK2R, CCK2-R, CCK-B, CCK-B receptor, CCK-BR, CCKRB, cholecystokinin B receptor, Cholecystokinin-2 receptor, cholecystokinin-B receptor/gastrin receptor, GASR, gastrin receptor, gastrin/cholecystokinin type B receptor | |
Goat | |
48 kDa | |
0.1 mg | |
Primary | |
Cross reacts with Human and Rat. Expected from sequence similarity: Mouse and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/ml, Immunohistochemistry 4-6 ug/ml, Immunohistochemistry-Paraffin 4-6 ug/ml, Peptide ELISA Detection limit 1:32000 | |
P32239 | |
CCKBR | |
Peptide with sequence C-FDGDSDSDSQSRVRNQ corresponding to internal region according to NP_795344.1. | |
Affinity Purified | |
RUO | |
887 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction