Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cholera Haemolysin Antibody (2024/251), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062460AF532
Description
Cholera Haemolysin Monoclonal specifically detects Cholera Haemolysin in Bacteria samples. It is validated for ELISA.Specifications
Cholera Haemolysin | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
NB100-62460 is specific for Vibrio cholerae haemolysin (HlyA). HlyA is an extracellular matrix damaging protein with a molecular weight of 65 kDa that exists a water-soluble monomer and an oligomeric amphipathic protein. It belongs to a class of proteins known as pore-forming toxins for their ability to punch holes in plasma membranes. | |
Store at 4°C in the dark. |
ELISA | |
2024/251 | |
ELISA | |
HlyA, VCA0219 | |
Native organisms | |
Primary | |
Bacteria | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction