Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholera Haemolysin Antibody (2024/251) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199790 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017477 0.025 mg
NB10062460 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017477 Supplier Novus Biologicals Supplier No. NB100624600.025MG

Mouse Monoclonal Antibody

Cholera Haemolysin Monoclonal specifically detects Cholera Haemolysin in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Cholera Haemolysin
Applications ELISA
Classification Monoclonal
Clone 2024/251
Concentration 3.1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation PBS with 0.09% Sodium Azide
Host Species Mouse
Immunogen Native organisms
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-62460 is specific for Vibrio cholerae haemolysin (HlyA). HlyA is an extracellular matrix damaging protein with a molecular weight of 65 kDa that exists a water-soluble monomer and an oligomeric amphipathic protein. It belongs to a class of proteins known as pore-forming toxins for their ability to punch holes in plasma membranes.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.