Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholera Toxin Beta Antibody (23043) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017627 0.025 mg
NB10064675 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017627 Supplier Novus Biologicals Supplier No. NB100646750.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cholera Toxin Beta Monoclonal specifically detects Cholera Toxin Beta in Bacteria samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Cholera Toxin Beta
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry
Classification Monoclonal
Clone 23043
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS (pH 7.4) with 0.09% Sodium Azide
Gene Alias Cholera enterotoxin B chain, Cholera enterotoxin beta chain, Cholera enterotoxin gamma chain, Cholera enterotoxin subunit B, Cholera toxin B protein, Choleragenoid, CTX B, CTXB, TOX B, toxB, VC1456
Host Species Mouse
Immunogen Vibrio cholerae
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64675 detects the labile enterotoxin normally associated with V. cholera and reacts with an epitope on the B chain. No reactivity was observed with other species tested including various E. coli serotypes.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.