Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cholera Toxin Beta Antibody (23043), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064675FR
Description
Cholera Toxin Beta Monoclonal specifically detects Cholera Toxin Beta in Bacteria samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Cholera Toxin Beta | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
RUO | |
NB100-64675 detects the labile enterotoxin normally associated with V. cholera and reacts with an epitope on the B chain. No reactivity was observed with other species tested including various E. coli serotypes. | |
Store at 4C in the dark. | |
IgG1 |
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
23043 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
Cholera enterotoxin B chain, Cholera enterotoxin beta chain, Cholera enterotoxin gamma chain, Cholera enterotoxin subunit B, Cholera toxin B protein, Choleragenoid, CTX B, CTXB, TOX B, toxB, VC1456 | |
Vibrio cholerae | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction