Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cholera Toxin beta Monoclonal Antibody (A68G)
GREENER_CHOICE

Catalog No. PIMA518189
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518189 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518189 Supplier Invitrogen™ Supplier No. MA518189
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with the B subunit of intact cholera toxin. Purity is >90%. Recommended pairs for ELISA: MA5-18189 (capture) and MA5-18188 or MA5-18190 (conjugated, detection).

Cholera toxin (sometimes abbreviated to CTX, Ctx, or CT) is a protein complex secreted by the bacterium Vibrio cholerae. CTX is the causative agent of cholera infection. The alpha subunit contains two chains, A1 and A2, linked by a disulfide bridge. The alpha subunit activates the adenylate cyclase enzyme in cells of the intestinal mucosa leading to increased levels of intracellular cAMP. The beta subunit of cholera toxin binds to a GM1-ganglioside receptor which is widely accepted to initiate toxin action by triggering uptake and delivery of the toxin alpha subunit into cells. The holotoxin consists of a pentameric ring of beta subunits whose central pore is occupied by the alpha subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cholera Toxin beta
Applications ELISA
Classification Monoclonal
Clone A68G
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias CTB; CTX; VC1456
Host Species Mouse
Immunogen Cholera toxin native protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.