Learn More
Invitrogen™ Cholera Toxin beta Monoclonal Antibody (F5J)

Description
This antibody reacts with the B subunit of intact cholera toxin. Purity is >90%. Recommended pairs for ELISA: MA5-18189 (capture) and MA5-18188 (conjugated, detection).
Specifications
Specifications
| Antigen | Cholera Toxin beta |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | F5J |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | CTB; CTX; VC1456 |
| Host Species | Mouse |
| Immunogen | Cholera toxin native protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.