Learn More
Invitrogen™ Cholera Toxin beta Polyclonal Antibody

Description
The PA1-73187 antibody reacts with Cholera toxin beta subunit. PA1-73187 has been successfully used in ELISA applications. The PA1-73187 antibody was raised against purified Choleragenoid.
Specifications
Specifications
| Antigen | Cholera Toxin beta |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 5.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | CTB; CTX; VC1456 |
| Host Species | Rabbit |
| Immunogen | Purified Choleragenoid. |
| Purification Method | IgG fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.