Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abbott™ Are You At Risk Patient Education Pamphlet
Click to view available options
Format:
Pamphlet
Specifications
Specifications
Format | Pamphlet |
Product Type | Patient Education Pamphlet |
For Use With (Application) | Education for patients regarding NCEP ATP III guidelines, explanations of cholesterol and diabetes test results and promotion of testing event |
Subject | Biology |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction