Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cholesterol
SCI_ED

Catalog No. s25677 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
Packaging:
Solid

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 57-88-5
Molecular Formula C27H46O
Molecular Weight (g/mol) 386.664
InChI Key HVYWMOMLDIMFJA-DPAQBDIFSA-N
Synonym cholesterol, cholesterin, cholest-5-en-3beta-ol, cholesteryl alcohol, cholestrin, cordulan, dusoline, dusoran, provitamin d, cholesterine
PubChem CID 5997
ChEBI CHEBI:16113
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C

Specifications

Density 1.067g/mL
Packaging Solid
Linear Formula C27H46O
Quantity 25 g
Formula Weight 385.31
Physical Form Solid
Chemical Name or Material Cholesterol
ShelfLife 2 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.