Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Cholesterol Dry Powder Reagents

Catalog No. 23666194 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
50 mL
600 mL
4 x 500 mL
Form:
Dry Powder
Liquid
Formulation:
4-Aminoantipyrine 0.25mM, Cholesterol Esterase >150U/L, Cholesterol Oxidase >150U/L, Peroxidase >1500U/L, Phenol >15mM, pH 6.8 Phosphate Buffer, non-reactive Stabilizers and preservatives
4-Aminoantipyrine 0.6mM, Sodium Cholate 8mM, Cholesterol Esterase <150μ/L, Cholesterol Oxidase <200μ/L, Horseradish Peroxidase 1000μ/L, p-Hydroxybenzene Sulfonate 20mM, Surfactants, Buffer pH 7.2, non-
pH:
6.7
7.2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Formulation pH
23-666-194 50 mL Liquid 4-Aminoantipyrine 0.25mM, Cholesterol Esterase >150U/L, Cholesterol Oxidase >150U/L, Peroxidase >1500U/L, Phenol >15mM, pH 6.8 Phosphate Buffer, non-reactive Stabilizers and preservatives 6.7
23-666-205 4 x 500 mL Dry Powder 4-Aminoantipyrine 0.6mM, Sodium Cholate 8mM, Cholesterol Esterase <150μ/L, Cholesterol Oxidase <200μ/L, Horseradish Peroxidase 1000μ/L, p-Hydroxybenzene Sulfonate 20mM, Surfactants, Buffer pH 7.2, non- 7.2
23-666-196 600 mL Liquid 4-Aminoantipyrine 0.25mM, Cholesterol Esterase >150U/L, Cholesterol Oxidase >150U/L, Peroxidase >1500U/L, Phenol >15mM, pH 6.8 Phosphate Buffer, non-reactive Stabilizers and preservatives 6.7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-666-194 Supplier Pointe Scientific Supplier No. HC710600

Includes

4-Aminoantipyrine 0.6mM, Sodium Cholate 8.0mM, Cholesterol Esterase ≥150u/L, Cholesterol Oxidase ≥200u/L, Horseradish Peroxidase 1000u/L, ρ-Hydroxybenzene Sulfonate 20mM, Surfactant, Buffer, pH 7.2, non-reactive stabilizers and fillers with Sodium Azide (.01%)

Quantify the total cholesterol in serum

  • Method: Enzymatic, Endpoint
  • Format: Dry powder
  • Wavelength: 520nm
  • Linearity: 500mg/dL
  • Expected Values: <200mg/dL (desirable cholesterol); 200-239mg/dL (borderline-high); >240mg/dL (high)
  • Reconstituted stability: 60 days at 2 to 8°C

For the quantitative determination of Total Cholesterol in serum

Specifications

Certifications/Compliance CE Marked
Type Cholesterol Reagent Set
Formulation 4-Aminoantipyrine 0.25mM, Cholesterol Esterase >150U/L, Cholesterol Oxidase >150U/L, Peroxidase >1500U/L, Phenol >15mM, pH 6.8 Phosphate Buffer, non-reactive Stabilizers and preservatives
For Use With (Application) Quantitative determination of total Cholesterol in Serum
pH 6.7
Quantity 50 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.