Learn More
Description
Includes
4-Aminoantipyrine 0.6mM, Sodium Cholate 8.0mM, Cholesterol Esterase ≥150u/L, Cholesterol Oxidase ≥200u/L, Horseradish Peroxidase 1000u/L, ρ-Hydroxybenzene Sulfonate 20mM, Surfactant, Buffer, pH 7.2, non-reactive stabilizers and fillers with Sodium Azide (.01%)
- Method: Enzymatic, Endpoint
- Format: Dry powder
- Wavelength: 520nm
- Linearity: 500mg/dL
- Expected Values: <200mg/dL (desirable cholesterol); 200-239mg/dL (borderline-high); >240mg/dL (high)
- Reconstituted stability: 60 days at 2 to 8°C
For the quantitative determination of Total Cholesterol in serum
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Cholesterol Reagent Set |
| Formulation | 4-Aminoantipyrine 0.25mM, Cholesterol Esterase >150U/L, Cholesterol Oxidase >150U/L, Peroxidase >1500U/L, Phenol >15mM, pH 6.8 Phosphate Buffer, non-reactive Stabilizers and preservatives |
| For Use With (Application) | Quantitative determination of total Cholesterol in Serum |
| pH | 6.7 |
| Quantity | 600 mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.