Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesterol, NF, 95-102%, Spectrum™ Chemical
SDP

Catalog No. 18601653 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

CH135, 57-88-5, C27H46O

Spectrum™ Chemical Cholesterol, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-88-5
Molecular Formula C27H46O
Molecular Weight (g/mol) 386.66
InChI Key HVYWMOMLDIMFJA-DPAQBDIFSA-N
IUPAC Name (1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.3%
Melting Range 147-150°C
Packaging Amber Glass Bottle
Quantity 125 g
Specific Rotation -34° to -38°
Percent Purity 95-102%
Grade NF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.