Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesteryl chloroformate, Thermo Scientific Chemicals

Catalog No. p-7044139
Click to view available options
Quantity:
25 g
100 g

Catalog No. AAL0480114

Supplier: Thermo Scientific Chemicals L0480114

Only null left
Add to Cart

Cholesteryl chloroformate is used in the preparation of hydrophobized chitosan oligosaccharide, which finds application as an efficient gene carrier. It acts as an initiator in the polymerization of methyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cholesteryl chloroformate is used in the preparation of hydrophobized chitosan oligosaccharide, which finds application as an efficient gene carrier. It acts as an initiator in the polymerization of methyl methacrylate.

Solubility
Soluble in methylene chloride, chloroform, acetone, toluene and benzene. Slightly soluble in methanol and ethanol.

Notes
Moisture sensitive. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7144-08-3
MDL Number MFCD00003633

Specifications

Melting Point 116°C to 120°C
Odor Pungent
Molecular Formula C5H11NO2
Quantity 25 g
UN Number UN3261
Beilstein 3223154
Sensitivity Moisture sensitive
Synonym n,n-dimethylglycine methyl ester, methyl 2-dimethylamino acetate, glycine, n,n-dimethyl-, methyl ester, methyl dimethylamino acetate, methyl n,n-dimethylglycinate, acmc-2097kq, dimethylglycine methyl ester, ch3 2nch2cooch3, dimethylaminoessigsauremethylester
Solubility Information Soluble in methylene chloride,chloroform,acetone,toluene and benzene. Slightly soluble in methanol and ethanol.
InChI Key LRZFEBJUJIQVDQ-UHFFFAOYSA-N
SMILES CN(C)CC(=O)OC
Optical Rotation -29° (c = 5 in Chloroform)
IUPAC Name methyl 2-(dimethylamino)acetate
Molecular Weight (g/mol) 117.15
PubChem CID 81569
Formula Weight 449.12
Chemical Name or Material Cholesteryl chloroformate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 230-447-9
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.