Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesteryl Hydrogen Succinate, MP Biomedicals™

Catalog No. 0210137980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-101-37980 100 g
ICN10137905 5 g
ICN10137925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-101-37980 Supplier MP Biomedicals, Inc Supplier No. 0210137980
Add to Cart
Add to Cart

Cholesteryl Hydrogen Succinate is a cholesterol derivative that is used as a detergent.

  • Dicarboxylic acid monoester resulting from the condensation of the cholesterol hydroxy group with a carboxy group of succinic acid.
  • Sometimes used as a replacement for cholesterol in protein crystallography.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1510-21-0
Molecular Formula C31H50O4
Molecular Weight (g/mol) 486.74
InChI Key WLNARFZDISHUGS-MIXBDBMTSA-N
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(=O)CCC(O)=O

Specifications

Quantity 100 g
IUPAC Name 4-{[(1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl]oxy}-4-oxobutanoic acid
Physical Form Powder
Chemical Name or Material Cholesteryl hydrogen succinate
Recommended Storage 15 to 30°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.