Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholestyramine Resin, USP, Spectrum™ Chemical
SDP

Catalog No. 18601661 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail

C1734, 11041-12-6

Spectrum™ Chemical Cholestyramine Resin, USP is a cholesterol lowering agent. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 11041-12-6
Molecular Formula C21H30ClN
Molecular Weight (g/mol) 331.93
InChI Key KNDHRUPPBXRELB-UHFFFAOYNA-M
IUPAC Name 4-[3-(4-ethylphenyl)butyl]-N,N,N-trimethylanilinium chloride
SMILES [Cl-].CCC1=CC=C(C=C1)C(C)CCC1=CC=C(C=C1)[N+](C)(C)C

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 12%
Packaging Amber Glass Bottle
Quantity 500 g
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.