Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-7081331
Click to view available options
Quantity:
25 g
100 g
500 g

Cholic acid is used as a preservative for food. It is widely used as an intermediate in the manufacture of pharmaceuticals. It finds use in biological research. It helps in detergents to soften biological fats. It helps in preventing and melting gallstones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cholic acid is used as a preservative for food. It is widely used as an intermediate in the manufacture of pharmaceuticals. It finds use in biological research. It helps in detergents to soften biological fats. It helps in preventing and melting gallstones.

Solubility
Soluble in alcohols, acetone, dimethylsulfoxide, methanol. Insoluble in water.

Notes
Stable. Incompatible with other materials such as strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-25-4
Molecular Formula C24H40O5
Molecular Weight (g/mol) 408.579
MDL Number MFCD00003672
InChI Key BHQCQFFYRZLCQQ-OELDTZBJSA-N
Synonym cholic acid, cholalic acid, cholate, cholalin, colalin, cholsaeure, cholbam, cholicacid, cholic acid, 5beta, 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanic acid
PubChem CID 221493
ChEBI CHEBI:16359
IUPAC Name (4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES CC(CCC(=O)O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Melting Point 199°C to 202°C
Quantity 25 g
Beilstein 2822009
Merck Index 14,2203
Solubility Information Soluble in alcohols,acetone,dimethylsulfoxide,methanol. Insoluble in water.
Optical Rotation +35 ± 2° (c=0.6 in ethanol)
Formula Weight 408.58
Percent Purity ≥98%
Chemical Name or Material Cholic acid
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 201-337-8
RTECSNumber FZ9350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.