Learn More
Cholic acid sodium salt, Thermo Scientific Chemicals
Description
Cholic acid sodium salt is a bio-surfactant which can be used as a reducing reagent in aqueous solution. It is also used as a non-denaturing detergent used for extraction of membrane proteins. Sodium cholate hydrate has been used in a study to assess the effect of phospholipid formulation and surfactants on the dermal delivery of hydrophilic drugs from elastic liposomes. It has also been used in a study to investigate the elastic deformable vesicles, transfersomes, for transdermal drug delivery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cholic acid sodium salt is a bio-surfactant which can be used as a reducing reagent in aqueous solution. It is also used as a non-denaturing detergent used for extraction of membrane proteins. Sodium cholate hydrate has been used in a study to assess the effect of phospholipid formulation and surfactants on the dermal delivery of hydrophilic drugs from elastic liposomes. It has also been used in a study to investigate the elastic deformable vesicles, transfersomes, for transdermal drug delivery.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
 
            Specifications
Specifications
| Color | White | 
| Quantity | 25 g | 
| Beilstein | 3582354 | 
| Merck Index | 14,2203 | 
| Solubility Information | It is soluble in water. | 
| Formula Weight | 430.6 | 
| Physical Form | Powder | 
| Chemical Name or Material | Cholic acid sodium salt | 
Safety and Handling
| TSCA | Yes | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
 
                                