Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholic Acid Sodium Salt, ≥97% (dry basis), Ultrapure

Catalog No. AAJ1363022
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
Packaging:
Plastic bottle

CAS: 361-09-1 | C24H39NaO5 | 430.561 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 361-09-1
Molecular Formula C24H39NaO5
Molecular Weight (g/mol) 430.561
MDL Number MFCD00150749
InChI Key NRHMKIHPTBHXPF-RLWQFIAZSA-M
Synonym sodium cholate, sodium 3,7,12-trihydroxycholan-24-oate, sodium cholate, reagent, ksc575s0b, 3,7,12-trihydroxycholan-24-oic acid sodium salt
PubChem CID 57349315
IUPAC Name sodium;(4 R)-4-[(8 R,9 S,10 S,13 R,14 S,17 R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1 H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES CC(CCC(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+]

Specifications

Physical Form Powder
Quantity 100 g
Beilstein 3582354
Formula Weight 430.55
Percent Purity ≥98%
Grade Ultrapure
Packaging Plastic bottle
Assay Dry basis
Chemical Name or Material Cholic Acid Sodium Salt
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-643-5
RTECSNumber FZ9600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.