Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Choline Acetyltransferase/ChAT Rabbit anti-Human, Clone: 1I4V0, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | Choline Acetyltransferase/ChAT |
---|---|
Clone | 1I4V0 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Choline Acetyltransferase/ChAT Monoclonal antibody specifically detects Choline Acetyltransferase/ChAT in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Choline Acetyltransferase/ChAT | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cellular Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1103 | |
IgG | |
Affinity purified |
1I4V0 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
acetyl CoA:choline O-acetyltransferase, ChAT, CHOACTase, Choline acetylase, choline acetyltransferase, choline O-acetyltransferase, CMS1A, CMS1A2, EC 2.3.1, EC 2.3.1.6 | |
A synthetic peptide corresponding to a sequence within amino acids 649-748 of human Choline Acetyltransferase/ChAT (P28329). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title