Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Bitartrate Dietary Supplement, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
87-67-2
C9H19NO7
253.25
QWJSAWXRUVVRLH-LREBCSMRSA-M
(2-hydroxyethyl)trimethylazanium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
C[N+](C)(C)CCO.O[C@H]([C@@H](O)C([O-])=O)C(O)=O
87-67-2
C9H19NO7
253.25
QWJSAWXRUVVRLH-LREBCSMRSA-M
(2-hydroxyethyl)trimethylazanium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
C[N+](C)(C)CCO.O[C@H]([C@@H](O)C([O-])=O)C(O)=O

C2052, 87-67-2, C9H19NO7

Spectrum™ Choline Bitartrate, USP Dietary Supplement is an essential nutrient and typically grouped with B-complex vitamins. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 50 kg
Ignition Residue 0.1%
Optical Rotation +17.5 to +18.5°
Packaging Fiber Drum
CAS Max % 100%
Percent Purity 99-100.5%