Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Bitartrate, FCC, 98%, Spectrum™ Chemical
SDP

Catalog No. 18601666
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

C1228, 87-67-2, C9H19NO7

Spectrum™ Chemical Choline Bitartrate, FCC is an essential nutrient and typically grouped with B-complex vitamins. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 87-67-2
Molecular Formula C9H19NO7
Molecular Weight (g/mol) 253.25
InChI Key QWJSAWXRUVVRLH-LREBCSMRSA-M
IUPAC Name (2-hydroxyethyl)trimethylazanium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
SMILES C[N+](C)(C)CCO.O[C@H]([C@@H](O)C([O-])=O)C(O)=O

Specifications

CAS Max % 100%
Quantity 2.5 kg
Percent Purity 98%
Grade FCC
Ignition Residue 0.1%
Optical Rotation +17.5 to +18.5°
Packaging Poly Bottle
Water 0.5%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.