Learn More
Choline chloride, 98+%
Description
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.
Solubility
Soluble in water, ethanol, methanol and alcohol.
Notes
Incompatible with strong oxidizing agents, strong bases, strong acids, alkalies and alloys.
Specifications
Specifications
| Melting Point | ∼305°C (decomposition) |
| Assay Percent Range | ≥98% |
| Quantity | 2500 g |
| Beilstein | 3563126 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,2206 |
| Solubility Information | Soluble in water,ethanol,methanol and alcohol. |
| Formula Weight | 139.63 |
| Percent Purity | ≥98% |
| Physical Form | Solid |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 200-655-4 |
| RTECSNumber | KH2975000 |
| TSCA | Yes |
| storageNote1 | Hygroscopic |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.