Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Chloride, MP Biomedicals

Catalog No. mp021013865
Click to view available options
Quantity:
500 mg
100 g

  • Choline chloride is used as a substrate for choline acetyl transferase

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
PubChem CID 6209
ChEBI CHEBI:133341
IUPAC Name (2-hydroxyethyl)trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

Melting Point 302°C to 305°C (decomposition)(literature)
Color White
Linear Formula (CH3)3N(Cl)CH2CH2OH
Quantity 500 mg
Solubility Information Slightly soluble in water (10 % aq. Soln - Clear and Colorless).
Formula Weight 139.6
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material CHOLINE CHLORIDE
Recommended Storage Store at Room Temperature (15-30°C),dessicate.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.