Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Choline Deficient Diet, Modified, Powder

Choline deficient diet, modified, trace minerals and fiber. Powder form.

$707.00 - $2683.00

Specifications

Product Type Animal Diet
Autoclavable Not Autoclavable
Disposable Single-use
Format Powder
Sterility Non-sterile
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP296020910
View Documents
MP Biomedicals
0296020910
10 kg
Each for $707.00
Only null left
Add to Cart
 
MP296020920
View Documents
MP Biomedicals
0296020920
20 kg
Each for $1,234.00
Only null left
Add to Cart
 
MP296020950
View Documents
MP Biomedicals
0296020950
50 kg
Each for $2,683.00
Only null left
Add to Cart
 
Description

Description

  • Choline deficient diet, modified is used for the study characterisation of a stereotypical cellular and extracellular adult liver progenitor cell niche in rodents and diseased human liver
  • Contains trace minerals and fiber
Specifications

Specifications

Animal Diet
Single-use
Non-sterile
1 ft.
Not Autoclavable
Powder
Refrigerated
0.3048 m
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.