Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Transporter Mouse anti-Mouse, Clone: 62-2E8, MilliporeSigma™

Catalog No. MAB5514I100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5514I100 100 μL
MAB5514I25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB5514I100 Supplier MilliporeSigma Supplier No. MAB5514I100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Choline Transporter Monoclonal antibody specifically detects Choline Transporter in Mouse samples. It is validated for Immunohistochemistry (Paraffin), ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Choline Transporter
Applications Immunohistochemistry (Paraffin), ELISA
Classification Monoclonal
Clone 62-2E8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 80 amino acids from the C-terminal region of human choline transporter.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.