Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chondroitin sulfate A sodium salt, 90%
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Chondroitin sulfate is a sulfated (in different position) glycosaminoglycan (GAG).Composed of repeating and alternating disaccharide units of D-glucuronic acid and N-acetylgalactosamine. In chondroitin sulfate A, sulfate is on carbon 4 of the N-acetylgalactosamine (GalNAc) sugar.
General Description
- Chondroitin sulfate A is characterized by a N-acetylgalactosamine molecule with a sulfated carbon-4
- Chondroitin sulfate A is found in the extracellular matrix and is an important structural polysaccharide
- Chondroitin sulfates play roles in cellular development and the formation of tissue barriers throughout the body
Application
- Chondroitin sulfate A is useful for in vitro study of nutrition and cellular-level repair
- Chondroitin sulfate A can be used in the laboratory study of cell signaling, tissue growth, and repair

Specifications
Specifications
Chemical Name or Material | Chondroitin sulfate A sodium salt |
Color | White |
Quantity | 25 g |
MDL Number | MFCD00130792 |
Solubility Information | Insoluble in water. |
Percent Purity | 90% |
Physical Form | Powder |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction