Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chondroitin Sulfate (NG2) Mouse anti-Human, PE, Clone: 7.1, BD Phrmingen™

Catalog No. BDB569962 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569962 0.1 mg
BDB569963 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569962 Supplier BD Biosciences Supplier No. 569962
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chondroitin Sulfate (NG2) Monoclonal antibody specifically detects Chondroitin Sulfate (NG2) in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chondroitin Sulfate (NG2)
Applications Flow Cytometry
Classification Monoclonal
Clone 7.1
Concentration 0.2 mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CSPG4; HMW-MAA; MCSP; MCSPG; NG2
Host Species Mouse
Immunogen Human BM stromal cell line established by infection with SV-40
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1464
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.