Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chondrolectin Antibody (25G6), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259424DL594
This item is not returnable.
View return policy
Description
Chondrolectin Monoclonal antibody specifically detects Chondrolectin in Human samples. It is validated for Western Blot, ELISASpecifications
Chondrolectin | |
Monoclonal | |
DyLight 594 | |
C21orf68, chondrolectin, chromosome 21 open reading frame 68, FLJ12627, MT75, PRED12, Transmembrane protein MT75 | |
Recombinant human Chondrolectin (22-216aa) purified from E. coli | |
0.1 mL | |
Developmental Biology | |
140578 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, ELISA | |
25G6 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction