Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Chondrolectin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569993
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 92%; Horse: 79%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 93%.
This gene encodes a type I membrane protein with a carbohydrate recognition domain characteristic of C-type lectins in its extracellular portion. In other proteins, this domain is involved in endocytosis of glycoproteins and exogenous sugar-bearing pathogens. This protein localizes predominantly to the perinuclear region. Several transcript variants encoding a few different isoforms have been found for this gene.
Specifications
Chondrolectin | |
Polyclonal | |
Unconjugated | |
CHODL | |
3110074E07Rik; C21orf68; CHODL; Chondrolectin; FLJ12627; LOW QUALITY PROTEIN: chondrolectin; MT75; PRED12; Transmembrane protein MT75; UNQ872/PRO1890 | |
Rabbit | |
Affinity chromatography | |
RUO | |
246048 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9CXM0 | |
CHODL | |
synthetic peptide corresponding to a region of mouse Chondrolectin (aa 161-210). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction