Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chondrolectin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553574 Supplier Invitrogen™ Supplier No. PA553574
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GISDGDFWIG LWRNGDGQTS GACPDLYQWS DGSNSQYRNW YTDEPSCGSE KCVVMYHQPT ANPGLGGPYL YQWNDDRCNM KHNYICKYEP EINPTAPVEK PYLTNQPGDT HQNVVVTEA Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

This gene encodes a type I membrane protein with a carbohydrate recognition domain characteristic of C-type lectins in its extracellular portion. In other proteins, this domain is involved in endocytosis of glycoproteins and exogenous sugar-bearing pathogens. This protein localizes predominantly to the perinuclear region. Several transcript variants encoding a few different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chondrolectin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHODL
Gene Accession No. Q9H9P2
Gene Alias 3110074E07Rik; C21orf68; CHODL; Chondrolectin; FLJ12627; LOW QUALITY PROTEIN: chondrolectin; MT75; PRED12; Transmembrane protein MT75; UNQ872/PRO1890
Gene Symbols CHODL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Chondrolectin. Recombinant protein control fragment (Product #RP-90335).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140578
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.