Learn More
Description
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC21331 and based on accession number NM_715. The protein contains a C-terminal DDK tag. |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens leukocyte cell derived chemotaxin 1 (LECT1), transcript variant 1, mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Synonym | BRICD3, BRICHOS domain containing 3, CHM1chondromodulin I, ChM-I, CHMI, chondromodulin, chondromodulin-1, Chondromodulin-I, leukocyte cell derived chemotaxin 1, Leukocyte cell-derived chemotaxin 1, multiple myeloma tumor suppressor 1, MYETS1 |
| Protein State | Native |
| For Use With (Application) | Western Blot |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
