Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHOP/GADD153 Mouse anti-Human, Clone: 4F3G1, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 667411IG150UL

Description
GADD153 is a small nuclear protein that is capable of dimerizing with transcription factors C/EBP alpha and beta. Once dimerized, this complex inhibits the normal binding and function of C/EBP to classical binding sites. Inversely, the C/EBP GADD153 dimer gains binding activity to other non classical C/EBP stress related targets. Under normal cellular conditions this protein is not expressed in detectable levels, but is highly unregulated during times of cellular/ER stress. Examples of GADD153 inducing stress include: treatment with tunicamycin, nutrient starvation and reducing agents that interfere with the calcium flux across the ER membrane.Specifications
CHOP/GADD153 | |
Monoclonal | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P35638 | |
DDIT3 | |
CHOP; GADD153 Fusion Protein Ag7354 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot | |
4F3G1 | |
Unconjugated | |
DDIT3 | |
C/EBP homologous protein, C/EBP homologous protein 10, CEBPZ, CHOP, CHOP 10, CHOP10, DDIT 3, DDIT3, GADD153 | |
Mouse | |
Protein A | |
RUO | |
1649 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction