Learn More
CHORDC1 Monoclonal Antibody (3G8)

Description
CHORDC1 Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | CHORDC1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3G8 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CHORDC1 |
| Gene Accession No. | Q9UHD1 |
| Gene Alias | 1110001O09Rik; AA409036; chord domain-containing protein 1; CHORDC1; chordc1b; CHORD-containing protein 1; CHP1; CHP-1; cysteine and histidine rich domain containing 1; cysteine and histidine-rich domain (CHORD) containing 1; cysteine and histidine-rich domain (CHORD) containing 1b; cysteine and histidine-rich domain (CHORD)-containing 1; cysteine and histidine-rich domain (CHORD)-containing, zinc binding protein 1; cysteine and histidine-rich domain (CHORD)-containing, zinc-binding protein 1; cysteine and histidine-rich domain-containing protein 1; MGC137302 protein; morgana; Protein morgana; zgc:63894 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.