Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHORDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583531
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583531 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583531 Supplier Invitrogen™ Supplier No. PA583531
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKGWSCCKRR TTDFSDFLSI VGCTKGRHNS EKPPEPVKPE VKTTEKKELC ELKPKFQEHI IQAPKPVEAI Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.

The cysteine and histidine-rich domain (CHORD)-containing protein (CHORDC1) is a member of a highly conserved protein family that contains the plant protein RAR1 and the mammalian protein melusin. In mammals, CHORDC1 is an ADP-dependent HSP90-interacting protein, and this interaction is dependent on the ability of HSP90 to bind nucleotides. Recent experiments indicate that CHORDC1 mRNA is diurnally regulated in mouse hypothalamus, and that this regulation alters during development, suggesting that CHORDC1 may play a role in circadian mechanisms in the mammalian brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHORDC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHORDC1
Gene Accession No. D4A4T9, Q9D1P4, Q9UHD1
Gene Alias 1110001O09Rik; AA409036; chord domain-containing protein 1; CHORDC1; chordc1b; CHORD-containing protein 1; CHP1; CHP-1; cysteine and histidine rich domain containing 1; cysteine and histidine-rich domain (CHORD) containing 1; cysteine and histidine-rich domain (CHORD) containing 1b; cysteine and histidine-rich domain (CHORD)-containing 1; cysteine and histidine-rich domain (CHORD)-containing, zinc binding protein 1; cysteine and histidine-rich domain (CHORD)-containing, zinc-binding protein 1; cysteine and histidine-rich domain-containing protein 1; MGC137302 protein; morgana; Protein morgana; zgc:63894
Gene Symbols CHORDC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHORDC1. Recombinant protein control fragment (Product #RP-97299).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26973, 315447, 66917
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.