Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chordin Antibody (065), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289221AF532
Description
Chordin Monoclonal specifically detects Chordin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Chordin | |
Monoclonal | |
Alexa Fluor 532 | |
CHRD, MGC133038 | |
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Chordin. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
65 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
8646 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction