Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Chordin-like 1/CHRDL1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | Chordin-like 1/CHRDL1 |
---|---|
Clone | 263737 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Chordin-like 1/CHRDL1 Monoclonal specifically detects Chordin-like 1/CHRDL1 in Human samples. It is validated for Western Blot.Specifications
Chordin-like 1/CHRDL1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
NP_001137454 | |
91851 | |
Mouse myeloma cell line NS0-derived recombinant human Chordin-like 1/CHRDL1 Glu28-Cys457 Accession # NP_001137454 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Chordin-like 1/CHRDL1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Chordin-Like 2 is observed. |
263737 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CHL, chordin-like 1, chordin-like protein 1, CHRDL1, CRDL1, dA141H5.1, Neuralin 1, neuralin-1, Neurogenesin-1, NRLN1, NRLN1neuralin 1, ventroptin, Ventroptin alpha, VOPT | |
CHRDL1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title