Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chordin-like 2/CHRDL2 Antibody (272905), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB24481M
Description
Chordin-like 2/CHRDL2 Monoclonal antibody specifically detects Chordin-like 2/CHRDL2 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Chordin-like 2/CHRDL2 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
25884 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
272905 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
chordin-like 2, chordin-like protein 2 | |
Mouse myeloma cell line NS0-derived recombinant human Chordin-like 2/CHRDL2, Arg28-Thr451, Accession # NP_056239 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction