Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chordin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557414
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557414 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557414 Supplier Invitrogen™ Supplier No. PA557414
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PNTCFFEGQQ RPHGARWAPN YDPLCSLCTC QRRTVICDPV VCPPPSCPHP VQAPDQCCPV CPEKQDVRDL PGLPRSRDPG EGCYFDGDRS W Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

This gene encodes a secreted protein that dorsalizes early vertebrate embryonic tissues by binding to ventralizing TGF-beta-like bone morphogenetic proteins and sequestering them in latent complexes. The encoded protein may also have roles in organogenesis and during adulthood. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their full length sequence has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chordin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHRD
Gene Accession No. Q9H2X0
Gene Alias Chd; Chordin; Chrd; UNQ217/PRO243
Gene Symbols CHRD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Chordin. Recombinant protein control fragment (Product #RP-96297).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8646
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.