Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Chordin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568401
Description
Chordin Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a secreted protein that dorsalizes early vertebrate embryonic tissues by binding to ventralizing TGF-beta-like bone morphogenetic proteins and sequestering them in latent complexes. The encoded protein may also have roles in organogenesis and during adulthood. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their full length sequence has not been determined.
Specifications
| Chordin | |
| Polyclonal | |
| Unconjugated | |
| CHRD | |
| Chd; Chordin; Chrd; UNQ217/PRO243 | |
| Rabbit | |
| affinity chromatography | |
| RUO | |
| 8646 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9H2X0 | |
| CHRD | |
| Synthetic protein corresponding to residues near the C terminal of human CHRD. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction