Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chordin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109536
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109536 Supplier Invitrogen™ Supplier No. PA5109536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PALAPAKPGG PGRPRDPNTC FFEGQQRPHG ARWAPNYDPL CSLCTCQRRT VICDPVVCPP PSCPHPVQAP DQCCPVCPEK QDVRDLPGLP RSRDPGEGCY FDGDRSWRAA GTRWHPVVPP FGLIKCAVCT CKGGTGEVHC EKVQCPRLAC AQPVRVNPTD CCKQCPVGSG AHPQLGDPMQ ADGPRGCRFA GQWFPESQSW HPSVPPFGEM SCITCRCGAG VPHCERDDCS LPLSCGSGKE SRCCSRCTAH RRPAPETRTD PELEKEAEGS.

This gene encodes a secreted protein that dorsalizes early vertebrate embryonic tissues by binding to ventralizing TGF-beta-like bone morphogenetic proteins and sequestering them in latent complexes. The encoded protein may also have roles in organogenesis and during adulthood. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their full length sequence has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chordin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CHRD
Gene Accession No. Q63148, Q9Z0E2
Gene Alias Chd; Chordin; Chrd; UNQ217/PRO243
Gene Symbols CHRD
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 686-955 of human CHRD (NP_003732.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117275, 12667
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.