Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chorionic Gonadotropin alpha/beta (hCG) Antibody (HCGab/52), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311526JF549
Description
Chorionic Gonadotropin alpha/beta (hCG) Monoclonal antibody specifically detects Chorionic Gonadotropin alpha/beta (hCG) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Chorionic Gonadotropin alpha/beta (hCG) | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HCGab/52 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CGB;CGB3;CGB5;CGB7;CGB8;Choriogonadotropin subunit beta 3;Choriogonadotropin subunit beta 7;hCGB;LHB | |
Purified Chorionic Gonadotropin alpha/beta (hCG) protein | |
0.1 mL | |
Cancer | |
1082 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction